Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: key | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079132.4 | NP_523856.2 | 5 - 6 | 1243 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |