Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259596.2 | NP_001246525.1 | 2 - 3 | 334 | 70 | |
NM_001259597.2 | NP_001246526.1 | 2 - 3 | 356 | 70 | ||
NM_001259598.2 | NP_001246527.1 | 2 - 3 | 344 | 70 | ||
NM_079145.4 | NP_523869.1 | 1 - 2 | 449 | 70 | ||
NM_167799.3 | NP_728481.1 | 2 - 3 | 353 | 70 | ||
NM_167800.3 | NP_728482.1 | 2 - 3 | 341 | 70 | ||
GenBank mRNA | AF176011.1 | - | 1 - 2 | 411 | 70 | |
AY051973.1 | - | 1 - 2 | 411 | 70 | ||
BT125928.1 | - | 2 - 3 | 341 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |