Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259606.2 | NP_001246535.1 | 8 - 9 | 2146 | 95 | |
NM_079147.4 | NP_523871.1 | 8 - 9 | 1693 | 95 | ||
GenBank mRNA | AF109308.1 | - | 8 - 9 | 1654 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |