Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274313.1 | NP_001261242.1 | 4 - 5 | 2648 | 66 | |
NM_079155.3 | NP_523879.2 | 4 - 5 | 2651 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |