Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169855.2 | NP_001163326.1 | 11 - 12 | 3631 | 104 | |
NM_079163.4 | NP_523887.2 | 11 - 12 | 3649 | 104 | ||
NM_167927.3 | NP_728695.1 | 11 - 12 | 3586 | 104 | ||
GenBank mRNA | AY061475.1 | - | 11 - 12 | 3687 | 104 | |
BT030128.1 | - | 11 - 12 | 3610 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |