Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169855.2 | NP_001163326.1 | 6 - 7 | 1907 | 62 | |
NM_079163.4 | NP_523887.2 | 6 - 7 | 1925 | 62 | ||
NM_167927.3 | NP_728695.1 | 6 - 7 | 1862 | 62 | ||
GenBank mRNA | AY061475.1 | - | 6 - 7 | 1963 | 62 | |
BT030128.1 | - | 6 - 7 | 1886 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |