Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169855.2 | NP_001163326.1 | 8 - 9 | 3186 | 68 | |
NM_079163.4 | NP_523887.2 | 8 - 9 | 3204 | 68 | ||
NM_167927.3 | NP_728695.1 | 8 - 9 | 3141 | 68 | ||
GenBank mRNA | AY061475.1 | - | 8 - 9 | 3242 | 68 | |
BT030128.1 | - | 8 - 9 | 3165 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |