Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yellow-g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079164.3 | NP_523888.1 | 1 - 2 | 511 | 86 | |
GenBank mRNA | BT150079.1 | - | 1 - 2 | 511 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |