Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079186.4 | NP_523910.1 | 1 - 2 | 375 | 92 | |
GenBank mRNA | AF241222.1 | - | 1 - 2 | 353 | 92 | |
AY122211.1 | - | 1 - 2 | 353 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |