Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr64c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259681.1 | NP_001246610.1 | 7 - 8 | 1546 | 65 | |
NM_079189.4 | NP_523913.2 | 1 - 2 | 230 | 65 | ||
GenBank mRNA | BT132939.1 | - | 1 - 2 | 230 | 65 |
Gene Symbol: Gr64b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168049.2 | NP_728921.1 | 7 - 8 | 1546 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |