Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274479.1 | NP_001261408.1 | 3 - 4 | 458 | 68 | |
NM_079195.3 | NP_523919.1 | 3 - 4 | 458 | 68 | ||
GenBank mRNA | BT001597.1 | - | 3 - 4 | 455 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |