Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079204.2 | NP_523928.1 | 5 - 6 | 4388 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |