Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274511.1 | NP_001261440.1 | 5 - 6 | 1674 | 61 | |
NM_079209.3 | NP_523933.3 | 4 - 5 | 1565 | 61 | ||
NM_168115.2 | NP_729059.2 | 4 - 5 | 994 | 61 | ||
NM_168116.2 | NP_729060.2 | 4 - 5 | 1063 | 61 | ||
GenBank mRNA | BT133475.1 | - | 4 - 5 | 1542 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |