Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079211.3 | NP_523935.1 | 6 - 7 | 1826 | 78 | |
GenBank mRNA | AY119579.1 | - | 6 - 7 | 1764 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |