Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014565.2 | NP_001014565.2 | 3 - 4 | 3768 | 83 | |
NM_079218.3 | NP_523942.2 | 3 - 4 | 3768 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |