Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014565.2 | NP_001014565.2 | 4 - 5 | 3875 | 70 | |
NM_079218.3 | NP_523942.2 | 4 - 5 | 3875 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |