Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144422.2 | NP_001137894.2 | 8 - 9 | 2038 | 78 | |
NM_001316444.1 | NP_001303373.1 | 8 - 9 | 2038 | 78 | ||
NM_001316445.1 | NP_001303374.1 | 8 - 9 | 2038 | 78 | ||
NM_079221.3 | NP_523945.2 | 5 - 6 | 1076 | 78 | ||
NM_168156.2 | NP_729142.1 | 8 - 9 | 2038 | 78 | ||
GenBank mRNA | AY060316.1 | - | 5 - 6 | 1059 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |