Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144422.2 | NP_001137894.2 | 9 - 10 | 2252 | 110 | |
NM_001316444.1 | NP_001303373.1 | 9 - 10 | 2252 | 110 | ||
NM_001316445.1 | NP_001303374.1 | 9 - 10 | 2252 | 110 | ||
NM_079221.3 | NP_523945.2 | 6 - 7 | 1290 | 110 | ||
NM_168156.2 | NP_729142.1 | 9 - 10 | 2252 | 110 | ||
GenBank mRNA | AY060316.1 | - | 6 - 7 | 1273 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |