Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sfl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169883.3 | NP_001163354.1 | 4 - 5 | 3970 | 94 | |
NM_079222.5 | NP_523946.1 | 5 - 6 | 4007 | 94 | ||
GenBank mRNA | AF175689.2 | - | 5 - 6 | 3988 | 94 | |
BT132640.1 | - | 5 - 6 | 4004 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |