Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169884.2 | NP_001163355.2 | 1 | 3249 | 126 | |
NM_001300028.1 | NP_001286957.1 | 1 | 3249 | 126 | ||
NM_001316454.1 | NP_001303383.1 | 1 | 3249 | 126 | ||
NM_001316455.1 | NP_001303384.1 | 1 | 3249 | 126 | ||
NM_079224.5 | NP_523948.1 | 1 | 3249 | 126 | ||
GenBank mRNA | BT030760.1 | - | 1 | 3246 | 126 | |
M81959.1 | - | 1 | 2698 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |