Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: melt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014572.4 | NP_001014572.1 | 2 - 3 | 864 | 64 | |
NM_001274566.1 | NP_001261495.1 | 3 - 4 | 1150 | 64 | ||
NM_079229.4 | NP_523953.2 | 2 - 3 | 864 | 64 | ||
GenBank mRNA | BT029129.1 | - | 2 - 3 | 838 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |