Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079234.4 | NP_523958.2 | 7 - 8 | 1660 | 82 | |
GenBank mRNA | AY071599.1 | - | 7 - 8 | 1626 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |