Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274630.1 | NP_001261559.1 | 2 - 3 | 408 | 58 | |
NM_079244.3 | NP_523968.1 | 2 - 3 | 408 | 58 | ||
GenBank mRNA | AY051859.1 | - | 2 - 3 | 378 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |