Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dally | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259756.2 | NP_001246685.1 | 4 - 5 | 1745 | 63 | |
NM_079259.4 | NP_523983.1 | 4 - 5 | 1745 | 63 | ||
GenBank mRNA | BT011348.1 | - | 4 - 5 | 1744 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |