Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp66E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079261.5 | NP_523985.1 | 3 - 4 | 414 | 90 | |
NM_168298.1 | NP_729423.1 | 3 - 4 | 308 | 90 | ||
NM_168299.2 | NP_729424.1 | 3 - 4 | 440 | 90 | ||
GenBank mRNA | AF220044.1 | - | 3 - 4 | 392 | 90 | |
AY118391.1 | - | 3 - 4 | 308 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |