Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp66E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079261.5 | NP_523985.1 | 4 - 5 | 503 | 67 | |
NM_168298.1 | NP_729423.1 | 4 - 5 | 397 | 67 | ||
NM_168299.2 | NP_729424.1 | 4 - 5 | 529 | 67 | ||
GenBank mRNA | AF220044.1 | - | 4 - 5 | 481 | 67 | |
AY118391.1 | - | 4 - 5 | 397 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |