Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274686.1 | NP_001261615.1 | 6 - 7 | 2270 | 67 | |
NM_001274687.1 | NP_001261616.1 | 6 - 7 | 2270 | 67 | ||
NM_079267.4 | NP_523991.2 | 5 - 6 | 930 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |