Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144459.2 | NP_001137931.1 | 2 - 3 | 1921 | 78 | |
NM_079296.4 | NP_524020.2 | 3 - 4 | 2014 | 78 | ||
GenBank mRNA | BT025199.1 | - | 2 - 3 | 1865 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |