Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi3K68D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169949.2 | NP_001163420.1 | 11 - 12 | 4576 | 92 | |
NM_079304.4 | NP_524028.2 | 10 - 11 | 4925 | 92 | ||
NM_168475.3 | NP_729743.1 | 11 - 12 | 4624 | 92 | ||
GenBank mRNA | AY058587.1 | - | 10 - 11 | 4867 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |