Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi3K68D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169949.2 | NP_001163420.1 | 14 - 15 | 5365 | 99 | |
NM_079304.4 | NP_524028.2 | 13 - 14 | 5714 | 99 | ||
NM_168475.3 | NP_729743.1 | 14 - 15 | 5413 | 99 | ||
GenBank mRNA | AY058587.1 | - | 13 - 14 | 5656 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |