Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi3K68D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169949.2 | NP_001163420.1 | 3 - 4 | 1193 | 103 | |
NM_001274795.1 | NP_001261724.1 | 2 - 3 | 1542 | 103 | ||
NM_079304.4 | NP_524028.2 | 2 - 3 | 1542 | 103 | ||
NM_168475.3 | NP_729743.1 | 3 - 4 | 1241 | 103 | ||
NM_168476.3 | NP_729745.1 | 3 - 4 | 1241 | 103 | ||
GenBank mRNA | AY058587.1 | - | 2 - 3 | 1484 | 103 | |
AY061189.1 | - | 3 - 4 | 1236 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |