Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274851.1 | NP_001261780.1 | 3 - 4 | 840 | 84 | |
NM_001274852.1 | NP_001261781.1 | 3 - 4 | 878 | 84 | ||
NM_079325.3 | NP_524049.1 | 3 - 4 | 842 | 84 | ||
GenBank mRNA | AY061164.1 | - | 3 - 4 | 811 | 84 | |
L11013.1 | - | 3 - 4 | 842 | 84 | ||
L11328.1 | - | 3 - 4 | 842 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |