Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274851.1 | NP_001261780.1 | 4 - 5 | 1326 | 103 | |
NM_001274852.1 | NP_001261781.1 | 4 - 5 | 1364 | 103 | ||
NM_079325.3 | NP_524049.1 | 4 - 5 | 1328 | 103 | ||
GenBank mRNA | AY061164.1 | - | 4 - 5 | 1297 | 103 | |
L11013.1 | - | 4 - 5 | 1328 | 103 | ||
L11328.1 | - | 4 - 5 | 1328 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |