Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274851.1 | NP_001261780.1 | 5 - 6 | 1626 | 90 | |
NM_079325.3 | NP_524049.1 | 5 - 6 | 1628 | 90 | ||
GenBank mRNA | AY061164.1 | - | 5 - 6 | 1597 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |