Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bru3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202161.2 | NP_001189090.1 | 11 - 12 | 2452 | 88 | |
NM_001202162.2 | NP_001189091.1 | 10 - 11 | 2410 | 88 | ||
NM_001202163.2 | NP_001189092.1 | 11 - 12 | 2488 | 88 | ||
NM_001202164.2 | NP_001189093.1 | 9 - 10 | 2275 | 88 | ||
NM_001202165.2 | NP_001189094.1 | 10 - 11 | 2434 | 88 | ||
NM_001202166.4 | NP_001189095.1 | 11 - 12 | 2512 | 88 | ||
NM_001202167.2 | NP_001189096.1 | 10 - 11 | 2416 | 88 | ||
NM_001202168.2 | NP_001189097.1 | 10 - 11 | 2729 | 88 | ||
NM_001202169.2 | NP_001189098.1 | 9 - 10 | 2257 | 88 | ||
NM_001202170.2 | NP_001189099.1 | 9 - 10 | 2299 | 88 | ||
NM_001202171.2 | NP_001189100.1 | 10 - 11 | 2374 | 88 | ||
NM_001274878.1 | NP_001261807.1 | 13 - 14 | 2872 | 88 | ||
NM_001274879.1 | NP_001261808.1 | 13 - 14 | 2608 | 88 | ||
NM_001382115.1 | NP_001369049.1 | 11 - 12 | 2488 | 88 | ||
NM_001382116.1 | NP_001369050.1 | 11 - 12 | 2488 | 88 | ||
NM_001382117.1 | NP_001369051.1 | 12 - 13 | 2473 | 88 | ||
NM_079335.6 | NP_524059.4 | 10 - 11 | 2353 | 88 | ||
NM_168557.3 | NP_729923.1 | 12 - 13 | 2530 | 88 | ||
GenBank mRNA | AY060398.1 | - | 12 - 13 | 2530 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |