Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eig71Ea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079364.4 | NP_524088.2 | 2 - 3 | 279 | 75 | |
GenBank mRNA | BT126196.1 | - | 2 - 3 | 261 | 75 | |
KX531693.1 | - | 2 - 3 | 230 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |