Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNApol-delta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079375.3 | NP_524099.2 | 1 - 2 | 154 | 77 | |
GenBank mRNA | AY113526.1 | - | 1 - 2 | 118 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |