Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144487.2 | NP_001137959.1 | 2 - 3 | 537 | 61 | |
NM_001144488.2 | NP_001137960.1 | 2 - 3 | 530 | 61 | ||
NM_079381.3 | NP_524105.2 | 2 - 3 | 562 | 61 | ||
NM_168659.2 | NP_730153.1 | 2 - 3 | 559 | 61 | ||
GenBank mRNA | AF098864.1 | - | 2 - 3 | 535 | 61 | |
AY075375.1 | - | 2 - 3 | 562 | 61 | ||
BT015186.1 | - | 2 - 3 | 573 | 61 | ||
BT030407.1 | - | 1914 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |