Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079384.5 | NP_524108.1 | 1 - 2 | 419 | 62 | |
GenBank mRNA | BT150167.1 | - | 1 - 2 | 416 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |