Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079407.3 | NP_524131.1 | 4 - 5 | 1683 | 105 | |
GenBank mRNA | BT126369.1 | - | 4 - 5 | 1683 | 105 | |
X78349.1 | - | 1121 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |