Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eip75B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079409.3 | NP_524133.2 | 1 - 2 | 1797 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |