Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275160.1 | NP_001262089.1 | 2 - 3 | 318 | 63 | |
NM_079453.3 | NP_524177.1 | 2 - 3 | 318 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |