Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ac78C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259928.2 | NP_001246857.1 | 13 - 14 | 2856 | 66 | |
NM_001275208.1 | NP_001262137.1 | 19 - 20 | 4655 | 66 | ||
NM_001275209.1 | NP_001262138.1 | 20 - 21 | 4866 | 66 | ||
NM_001275210.1 | NP_001262139.1 | 20 - 21 | 4863 | 66 | ||
NM_079470.5 | NP_524194.3 | 20 - 21 | 4866 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |