Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ac78C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259928.2 | NP_001246857.1 | 14 - 15 | 3258 | 65 | |
NM_001275208.1 | NP_001262137.1 | 20 - 21 | 5057 | 65 | ||
NM_001275209.1 | NP_001262138.1 | 21 - 22 | 5268 | 65 | ||
NM_001275210.1 | NP_001262139.1 | 21 - 22 | 5265 | 65 | ||
NM_079470.5 | NP_524194.3 | 21 - 22 | 5268 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |