Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten-m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079491.3 | NP_524215.2 | 7 - 8 | 3251 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |