Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043156.3 | NP_001036621.1 | 2 - 3 | 781 | 88 | |
NM_001259961.2 | NP_001246890.1 | 2 - 3 | 781 | 88 | ||
NM_001259962.2 | NP_001246891.1 | 2 - 3 | 781 | 88 | ||
NM_001259964.2 | NP_001246893.1 | 2 - 3 | 781 | 88 | ||
NM_001275303.1 | NP_001262232.1 | 2 - 3 | 781 | 88 | ||
NM_079494.5 | NP_524218.4 | 2 - 3 | 781 | 88 | ||
GenBank mRNA | BT150396.1 | - | 2 - 3 | 599 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |