Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274003.1 | NP_001260932.1 | 2 - 3 | 605 | 61 | |
NM_136982.3 | NP_610826.1 | 1 - 2 | 480 | 61 | ||
GenBank mRNA | AY069605.1 | - | 1 - 2 | 461 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |