Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299442.1 | NP_001286371.1 | 5 - 6 | 1863 | 70 | |
NM_136991.4 | NP_610835.2 | 5 - 6 | 1863 | 70 | ||
GenBank mRNA | BT004493.1 | - | 5 - 6 | 1834 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |