Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259368.2 | NP_001246297.1 | 4 - 5 | 1715 | 98 | |
NM_001259369.2 | NP_001246298.1 | 4 - 5 | 1715 | 98 | ||
NM_137031.3 | NP_610875.1 | 4 - 5 | 1715 | 98 | ||
GenBank mRNA | AY051853.1 | - | 4 - 5 | 1637 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |