Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259374.2 | NP_001246303.1 | 4 - 5 | 998 | 61 | |
NM_001259375.2 | NP_001246304.1 | 4 - 5 | 1389 | 61 | ||
NM_137058.4 | NP_610902.1 | 4 - 5 | 1389 | 61 | ||
GenBank mRNA | AY069830.1 | - | 4 - 5 | 1709 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |